
Propanolamine

Synonym(s):
3-Amino-1-propanol, 3-Aminopropyl alcohol
Linear Formula:
HO(CH2)3NH2
CAS Number:
Molecular Weight:
75.11
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing