Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium iodate
Synonym(s):
Potassium iodate
Linear Formula:
KIO
3
CAS Number:
7758-05-6
Molecular Weight:
214.00
EC Number:
231-831-9
Products
Product Category
salts (5)
Brand
SAFC (1)
Sigma-Aldrich (4)
Supelco (4)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Reference Material (1)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
215929
Potassium iodate,
ACS reagent, 99.5%
Expand
View Pricing
1.05051
Potassium iodate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
207977
Potassium iodate,
reagent grade, ≥98%
Expand
View Pricing
1.02404
Potassium iodate,
volumetric standard, secondary reference material for iodometry, traceable to NIST SRM Certipur
®
Expand
View Pricing
438464
Potassium iodate,
99.995% trace metals basis
Expand
View Pricing
1.05050
Potassium iodate,
extra pure FCC
Expand
View Pricing
1.93662
Potassium iodate,
EMPLURA
®
Expand
View Pricing
60386
Potassium iodate,
reference material for titrimetry, certified by BAM, >99.5%
Expand
View Pricing
1.93262
Potassium iodate,
for analysis EMPARTA
®
ACS
Expand
View Pricing
24-4820
Potassium iodate,
SAJ first grade, ≥99.5%
Expand
View Pricing