Skip to Content
Merck
Partisil™ 10 ODS HPLC Column 10 μm particle size, L × I.D. 25 cm × 4.6 mm

Partisil 10 ODS HPLC Column

Partisil™ 10 ODS HPLC Column 10 μm particle size, L × I.D. 25 cm × 4.6 mm
Synonym(s):
Partisil HPLC Column

Products

Product Category
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1