
Pantoprazole Related Compound C

Synonym(s):
5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole
Empirical Formula (Hill Notation):
C8H6F2N2OS
CAS Number:
Molecular Weight:
216.21
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2