![Olaquindox VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/289/469/14349f4f-3ff2-4a8b-bf56-cc314b8173e4/640/14349f4f-3ff2-4a8b-bf56-cc314b8173e4.png)
Olaquindox
![Olaquindox VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/289/469/14349f4f-3ff2-4a8b-bf56-cc314b8173e4/640/14349f4f-3ff2-4a8b-bf56-cc314b8173e4.png)
Synonym(s):
N-(2-Hydroxyethyl)-3-methyl-2-quinoxalinecarboxamide 1,4-dioxide
Empirical Formula (Hill Notation):
C12H13N3O4
Products
Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing