Skip to Content
Merck
Norfentanyl-D5 oxalate solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Norfentanyl-D5 oxalate solution

Norfentanyl-D5 oxalate solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C16D5H17N2O5
CAS Number:
Molecular Weight:
327.39
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique