Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
′-Methylenebisacrylamide
Synonym(s):
BIS, Bis-acrylamide, N,N′-Methylenebisacrylamide
Linear Formula:
(H
2
C=CHCONH)
2
CH
2
CAS Number:
110-26-9
Molecular Weight:
154.17
EC Number:
203-750-9
Products
Brand
Sigma-Aldrich (5)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
M7279
N
,
N
′-Methylenebisacrylamide,
powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Expand
View Pricing
146072
N
,
N
′-Methylenebisacrylamide,
99%
Expand
View Pricing
M7256
N
,
N
′-Methylenebisacrylamide,
suitable for electrophoresis (after filtration or allowing insolubles to settle)
Expand
View Pricing
1.01546
N
,
N
′-Methylenebisacrylamide,
for electrophoresis, special grade for molecular biology
Expand
View Pricing
19-3180
N
,
N
′-Methylenebisacrylamide,
≥99.0%
Expand
View Pricing