
N-Methoxysuccinyl-Ala-Ala-Pro-Val p-nitroanilide

Synonym(s):
N-(Methoxysuccinyl)-L-alanyl-L-alanyl-L-prolyl-L-valine 4-nitroanilide, N-(Methoxysuccinyl)-Ala-Ala-Pro-Val 4-nitroanilide
Empirical Formula (Hill Notation):
C27H38N6O9
CAS Number:
Molecular Weight:
590.63
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing