
n-Butyl acetate

Synonym(s):
Butyl acetate
Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing