
Naproxen Related Compound G

Synonym(s):
(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (2R)-2-(6-Methoxy-2-naphthyl)propionic acid, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen
Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2