
Nandrolone decanoate

Synonym(s):
19-Nortestosterone 17-decanoate, 17β-Hydroxy-19-nor-4-androsten-3-one 17-decanoate, 17β-Hydroxy-4-estren-3-one 17-decanoate, 4-Estren-17β-ol-3-one 17-decanoate, Decadurabolin, Nandrolone decanoate
Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing