
N-Acetyl-DL-tryptophan

Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Amino Acid
Reference Material Type
Showing 1-4 of 4
Compare
Product Number
Description
Pricing