
Methyl Red solution

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing