
Methylphenidate hydrochloride

Synonym(s):
DL-threo-Methylphenidate hydrochloride
Empirical Formula (Hill Notation):
C14H19NO2 · HCl
CAS Number:
Molecular Weight:
269.77
Products
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2