
Methyl octanoate

Synonym(s):
Caprylic acid methyl ester, Methyl caprylate, Methyl octanoate, Octanoic acid methyl ester
Linear Formula:
CH3(CH2)6COOCH3
CAS Number:
Molecular Weight:
158.24
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique