
Methylglyoxal solution

Synonym(s):
Acetylformaldehyde, Pyruvaldehyde, Pyruvic aldehyde
Linear Formula:
CH3COCHO
CAS Number:
Molecular Weight:
72.06
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing