![Manganese(II) sulfate monohydrate EMPLURA®](/deepweb/assets/sigmaaldrich/product/structures/251/569/9ab9f3b3-0a9e-4235-a82b-a2cb21d54d0f/640/9ab9f3b3-0a9e-4235-a82b-a2cb21d54d0f.png)
Manganese(II) sulfate monohydrate
![Manganese(II) sulfate monohydrate EMPLURA®](/deepweb/assets/sigmaaldrich/product/structures/251/569/9ab9f3b3-0a9e-4235-a82b-a2cb21d54d0f/640/9ab9f3b3-0a9e-4235-a82b-a2cb21d54d0f.png)
Synonym(s):
Manganese(II) sulfate monohydrate
Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing