Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Maleic anhydride
Synonym(s):
2,5-Furandione, Maleic anhydride
Empirical Formula (Hill Notation):
C
4
H
2
O
3
CAS Number:
108-31-6
Molecular Weight:
98.06
EC Number:
203-571-6
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
M188
Maleic anhydride,
99%
Expand
View Pricing
8.00408
Maleic anhydride,
for synthesis
Expand
View Pricing
89529
Maleic anhydride,
analytical standard
Expand
View Pricing
63200
Maleic anhydride,
puriss., ≥99.0% (NT)
Expand
View Pricing