
L-Threonine

Synonym(s):
(2S,3R)-2-Amino-3-hydroxybutyric acid, L-Threonine
Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing