![Loperamide hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/159/586/33cc802e-8426-42a2-9af4-1a977ce68d34/640/33cc802e-8426-42a2-9af4-1a977ce68d34.png)
Loperamide hydrochloride
![Loperamide hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/159/586/33cc802e-8426-42a2-9af4-1a977ce68d34/640/33cc802e-8426-42a2-9af4-1a977ce68d34.png)
Synonym(s):
4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutyramide hydrochloride
Empirical Formula (Hill Notation):
C29H33ClN2O2 · HCl
CAS Number:
Molecular Weight:
513.50
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing