Skip to Content
Merck
Iron(II) sulfate heptahydrate JIS special grade, 99.0-102.0%

Iron(II) sulfate heptahydrate

Iron(II) sulfate heptahydrate JIS special grade, 99.0-102.0%
Synonym(s):
Ferrous sulfate heptahydrate, Iron(II) sulfate heptahydrate
Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number: