
Iodomethane

Synonym(s):
Methyl iodide, Iodomethane
Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Special Grade
Showing 1-8 of 8
Compare
Product Number
Description
Pricing