
Hydroxyapatite

Synonym(s):
Calcium phosphate tribasic, Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate
Linear Formula:
[Ca5(OH)(PO4)3]x
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Showing 1-5 of 5
Compare
Product Number
Description
Pricing