
Hexanoyl-L-carnitine

Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium inner salt, (R)-Caproylcarnitine, n-Hexanoyl-L-carnitine, C6-Carnitine
Empirical Formula (Hill Notation):
C13H25NO4
CAS Number:
Molecular Weight:
259.34
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing