Skip to Content
Merck
Harmine phyproof® Reference Substance

Harmine

Harmine phyproof® Reference Substance
Synonym(s):
7-Methoxy-1-methyl-9H-pyrido[3,4-b]indole, Banisterine, Leucoharmine, Telepathine, Yageine
Empirical Formula (Hill Notation):
C13H12N2O
CAS Number:
Molecular Weight:
212.25

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing