Skip to Content
Merck
Gliclazide British Pharmacopoeia (BP) Reference Standard

Gliclazide

Gliclazide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1-(3-Azabicyclo[3.3.0]oct-3-yl)-3-p-tolylsulphonylurea
Empirical Formula (Hill Notation):
C15H21N3O3S
CAS Number:
Molecular Weight:
323.41
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3