Skip to Content
Merck
Ethyl formate SAJ special grade, ≥97.0%

Ethyl formate

Ethyl formate SAJ special grade, ≥97.0%
Synonym(s):
Formic acid ethyl ester
Linear Formula:
HCOOC2H5
CAS Number:
Molecular Weight:
74.08
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique