
Eszopiclone

Synonym(s):
(+)-Eszopiclone, (+)-(5S)-6-(5-Chloropyridin-2-yl)-7-oxo-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate, (S)-Zopiclone
Empirical Formula (Hill Notation):
C17H17ClN6O3
CAS Number:
Molecular Weight:
388.81
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing