
Dynorphin A Porcine Fragment 1-13

Empirical Formula (Hill Notation):
C75H126N24O15
CAS Number:
Molecular Weight:
1603.95
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing