
Dutasteride for system suitability

Synonym(s):
Dutasteride, (5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost-1-ene-17-carboxamide
Empirical Formula (Hill Notation):
C27H30F6N2O2
CAS Number:
Molecular Weight:
528.53
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing