
Dimethylvinphos

Synonym(s):
2-Chloro-1-(2,4-dichlorophenyl)vinyl dimethyl phosphate, Dimethylvinfos
Empirical Formula (Hill Notation):
C10H10Cl3O4P
CAS Number:
Molecular Weight:
331.52
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing