
Dihydrotanshinone I

Synonym(s):
Dihydrotanshinone I, (-)-Dihydrotanshinone I, 15,16-Dihydrotanshinone I
Empirical Formula (Hill Notation):
C18H14O3
CAS Number:
Molecular Weight:
278.30
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing