Skip to Content
Merck
Diethylenetriaminepentaacetic acid suitable for suitability for complexometry, ≥99.0%

Diethylenetriaminepentaacetic acid

Diethylenetriaminepentaacetic acid suitable for suitability for complexometry, ≥99.0%
Synonym(s):
(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid, Diethylenetriaminepentaaceticacid
Linear Formula:
[(HOOCCH2)2NCH2CH2]2NCH2COOH
CAS Number:
Molecular Weight:
393.35
EC Number:

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Functional Group
Reaction Type
Special Grade
Technique