
Diethylamino Hydroxybenzoyl Hexyl Benzoate

Synonym(s):
Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate, Diethylamino hydroxybenzoyl hexyl benzoate
Empirical Formula (Hill Notation):
C24H31NO4
CAS Number:
Molecular Weight:
397.51
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing