![Desmethyl-pirimicarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/799/199/7c6cc8c8-749c-4da8-822b-d9641ef52ec7/640/7c6cc8c8-749c-4da8-822b-d9641ef52ec7.png)
Desmethyl-pirimicarb
![Desmethyl-pirimicarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/799/199/7c6cc8c8-749c-4da8-822b-d9641ef52ec7/640/7c6cc8c8-749c-4da8-822b-d9641ef52ec7.png)
Synonym(s):
5,6-Dimethyl-2-methylamino-4-pyrimidinyl dimethylcarbamate
Empirical Formula (Hill Notation):
C10H16N4O2
CAS Number:
Molecular Weight:
224.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing