
D-Camphor Impurity D

Synonym(s):
(−)-Fenchone, (−)-1,3,3-Trimethyl-2-norbornanone, (1R)-1,3,3-Trimethylbicyclo[2.2.1]heptan-2-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing