
Copper(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine

Synonym(s):
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine copper(II) salt
Empirical Formula (Hill Notation):
C80H88CuN8O8
CAS Number:
Molecular Weight:
1353.15
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing