
Colcemid

Synonym(s):
Demecolcine, N-Deacetyl-N-methylcolchicine, Colcemid
Empirical Formula (Hill Notation):
C21H25NO5
CAS Number:
Molecular Weight:
371.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Quality Segment
Sterility
Sterilization Method
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing