
Clonazepam Related Compound B

Synonym(s):
2-Amino-2′-chloro-5-nitrobenzophenone, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone
Empirical Formula (Hill Notation):
C13H9ClN2O3
CAS Number:
Molecular Weight:
276.68
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2