
cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester

Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2CH3
CAS Number:
Molecular Weight:
316.48
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing