![Butane-1,3-diol pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/227/105/5ce96bd8-fd69-4a74-8037-6efeb4e9f3a6/640/5ce96bd8-fd69-4a74-8037-6efeb4e9f3a6.png)
Butane-1,3-diol
![Butane-1,3-diol pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/227/105/5ce96bd8-fd69-4a74-8037-6efeb4e9f3a6/640/5ce96bd8-fd69-4a74-8037-6efeb4e9f3a6.png)
Synonym(s):
(±)-1,3-Butanediol, 1,3-Butylene glycol
Linear Formula:
CH3CH(OH)CH2CH2OH
CAS Number:
Molecular Weight:
90.12
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2