
Bromoacetic acid N-hydroxysuccinimide ester

Empirical Formula (Hill Notation):
C6H6BrNO4
CAS Number:
Molecular Weight:
236.02
Products
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing