
BRAND® BLAUBRAND® volumetric flask

Synonym(s):
BRAND® BLAUBRAND® volumetric flask, PP stopper, clear glass, BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
Products
Product Category
Brand
Manufacturer
Capacity (mL)
Cap
Glassware Class
Color
Package Size
Sterility
Style
Showing 1-12 of 12
Compare
Product Number
Description
Pricing