Skip to Content
Merck
Betaxolol hydrochloride European Pharmacopoeia (EP) Reference Standard

Betaxolol hydrochloride

Betaxolol hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
SL 75212
Empirical Formula (Hill Notation):
C18H29NO3 · HCl
CAS Number:
Molecular Weight:
343.89

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade