Skip to Content
Merck
Benzoic acid European Pharmacopoeia (EP) Reference Standard

Benzoic acid

Benzoic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Benzoic acid
Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-20 of 23
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2