Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium persulfate
Synonym(s):
AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER
Linear Formula:
(NH
4
)
2
S
2
O
8
CAS Number:
7727-54-0
Molecular Weight:
228.20
EC Number:
231-786-5
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (8)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
248614
Ammonium persulfate,
ACS reagent, ≥98.0%
Expand
View Pricing
A3678
Ammonium persulfate,
for molecular biology, suitable for electrophoresis, ≥98%
Expand
View Pricing
215589
Ammonium persulfate,
reagent grade, 98%
Expand
View Pricing
09913
Ammonium persulfate,
BioUltra, for molecular biology, ≥98.0% (RT)
Expand
View Pricing
A7460
Ammonium persulfate,
BioXtra, ≥98.0%
Expand
View Pricing
908932
Ammonium persulfate,
free-flowing, Redi-Dri
™
, ACS reagent, ≥98.0%
Expand
View Pricing
01-4910
Ammonium persulfate,
JIS special grade, ≥98.0%
Expand
View Pricing
911127
Ammonium persulfate,
free-flowing, Redi-Dri
™
, reagent grade, 98%
Expand
View Pricing
01-4920
Ammonium persulfate,
SAJ first grade, ≥95.0%
Expand
View Pricing