Skip to Content
BB
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
8-Hydroxyquinoline
Synonym(s):
8-Quinolinol, 8-Hydroxyquinoline, 8-Oxychinolin, Oxine
Empirical Formula (Hill Notation):
C
9
H
7
NO
CAS Number:
148-24-3
Molecular Weight:
145.16
EC Number:
205-711-1
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Feature
Formula Weight
Physical Form
Purity
Activity Spectrum
Agency
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
Reaction Type
Special Grade
ACS reagent (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
252565
8-Hydroxyquinoline,
ACS reagent, 98.5%
Expand
View Pricing
8.20261
8-Hydroxyquinoline,
for synthesis
Expand
View Pricing
1.07098
8-Hydroxyquinoline,
GR for analysis Reag. Ph Eur
Expand
View Pricing
H6878
8-Hydroxyquinoline,
Expand
View Pricing