4-(Diphenylamino)benzaldehyde
Synonym(s):
4-Formyltriphenylamine
Empirical Formula (Hill Notation):
C19H15NO
CAS Number:
Molecular Weight:
273.33
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing