![3,4-Bis(hydroxymethyl)furan 98%](/deepweb/assets/sigmaaldrich/product/structures/414/663/60b77fee-648b-4c56-a3b9-7499df3971f3/640/60b77fee-648b-4c56-a3b9-7499df3971f3.png)
3,4-Bis(hydroxymethyl)furan
![3,4-Bis(hydroxymethyl)furan 98%](/deepweb/assets/sigmaaldrich/product/structures/414/663/60b77fee-648b-4c56-a3b9-7499df3971f3/640/60b77fee-648b-4c56-a3b9-7499df3971f3.png)
Synonym(s):
Furan-3,4-dimethanol
Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing