Skip to Content
Merck
1-Octadecene analytical standard, ≥99.0% (GC)

1-Octadecene

1-Octadecene analytical standard, ≥99.0% (GC)
Synonym(s):
1-Octadecene
Linear Formula:
CH3(CH2)15CH=CH2
CAS Number:
Molecular Weight:
252.48
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5